Assay Details
Gene Symbol: LOC4329643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767619.2 | XP_015623105.1 | 10 - 11 | 1086 | 63 | |
XM_015767621.2 | XP_015623107.1 | 10 - 11 | 1071 | 63 | ||
GenBank mRNA | AB114856.1 | - | 10 - 11 | 1089 | 63 | |
AK100016.1 | - | 7 - 8 | 594 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
