Assay Details
| Gene Symbol: LOC4329643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767619.2 | XP_015623105.1 | 3 - 4 | 481 | 81 | |
| XM_015767621.2 | XP_015623107.1 | 3 - 4 | 466 | 81 | ||
| GenBank mRNA | AB114856.1 | - | 3 - 4 | 484 | 81 | |
More Information
Set Membership: |
|

LOC4329643
Genomic Map