Assay Details
| Gene Symbol: LOC4329696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769671.2 | XP_015625157.1 | 3 - 4 | 995 | 101 | |
| XM_015769672.2 | XP_015625158.1 | 2 - 3 | 140 | 101 | ||
| GenBank mRNA | AK243408.1 | - | 3 - 4 | 961 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769672.2 transcripts.
Set Membership: |
|

LOC4329696
Genomic Map