Assay Details
| Gene Symbol: LOC4329696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769671.2 | XP_015625157.1 | 9 - 10 | 1410 | 73 | |
| XM_015769672.2 | XP_015625158.1 | 8 - 9 | 555 | 73 | ||
| GenBank mRNA | AK243408.1 | - | 9 - 10 | 1376 | 73 | |
More Information
Set Membership: |
|

LOC4329696
Genomic Map