Assay Details
Gene Symbol: LOC4329935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769834.2 | XP_015625320.1 | 2 - 3 | 432 | 127 | |
XM_015769835.2 | XP_015625321.1 | 2 - 3 | 431 | 127 | ||
GenBank mRNA | AK073527.1 | - | 2 - 3 | 396 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769834.2, XM_015769835.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
