Assay Details
| Gene Symbol: LOC4329935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769834.2 | XP_015625320.1 | 7 - 8 | 2855 | 97 | |
| XM_015769835.2 | XP_015625321.1 | 7 - 8 | 2848 | 97 | ||
| GenBank mRNA | AK073527.1 | - | 7 - 8 | 2813 | 97 | |
More Information
Set Membership: |
|

LOC4329935
Genomic Map