Assay Details
| Gene Symbol: LOC4330051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768577.2 | XP_015624063.1 | 1 - 2 | 151 | 68 | |
| GenBank mRNA | AK121915.1 | - | 1 - 2 | 149 | 68 | |
More Information
Set Membership: |
|

LOC4330051
Genomic Map