Assay Details
| Gene Symbol: LOC4330348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771286.2 | XP_015626772.1 | 8 - 9 | 3118 | 99 | |
| GenBank mRNA | AK106462.1 | - | 3 - 4 | 1202 | 99 | |
More Information
Set Membership: |
|

LOC4330348
Genomic Map