Assay Details
| Gene Symbol: LOC4330621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771308.1 | XP_015626794.1 | 2 - 3 | 602 | 61 | |
| XM_015771309.1 | XP_015626795.1 | 2 - 3 | 600 | 61 | ||
| GenBank mRNA | AK241644.1 | - | 2 - 3 | 596 | 61 | |
| AY177702.1 | - | 2 - 3 | 261 | 61 | ||
| AY224482.1 | - | 2 - 3 | 261 | 61 | ||
More Information
Set Membership: |
|

LOC4330621
Genomic Map