Assay Details
Gene Symbol: LOC4330621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771308.1 | XP_015626794.1 | 3 - 4 | 660 | 93 | |
XM_015771309.1 | XP_015626795.1 | 3 - 4 | 658 | 93 | ||
GenBank mRNA | AK241644.1 | - | 3 - 4 | 654 | 93 | |
AY177702.1 | - | 3 - 4 | 319 | 93 | ||
AY224482.1 | - | 3 - 4 | 319 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
