Assay Details
Gene Symbol: LOC4330621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771309.1 | XP_015626795.1 | 7 - 8 | 1050 | 93 | |
GenBank mRNA | AK241644.1 | - | 7 - 8 | 1046 | 93 | |
AY177702.1 | - | 7 - 8 | 711 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
