Assay Details
Gene Symbol: LOC4331512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774266.2 | XP_015629752.1 | 5 - 6 | 974 | 102 | |
GenBank mRNA | AK109755.1 | - | 5 - 6 | 942 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
