Assay Details
Gene Symbol: LOC4331822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775390.2 | XP_015630876.1 | 1 - 2 | 677 | 63 | |
GenBank mRNA | AK107046.1 | - | 1 - 2 | 677 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
