Assay Details
Gene Symbol: LOC4331872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775124.1 | XP_015630610.1 | 6 - 7 | 875 | 92 | |
GenBank mRNA | AY345222.1 | - | 6 - 7 | 561 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
