Assay Details
| Gene Symbol: LOC4331978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777231.2 | XP_015632717.1 | 3 - 4 | 415 | 111 | |
| GenBank mRNA | AK121020.1 | - | 3 - 4 | 360 | 111 | |
| AY224468.1 | - | 3 - 4 | 232 | 111 | ||
More Information
Set Membership: |
|

LOC4331978
Genomic Map