Assay Details
Gene Symbol: LOC4331978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777231.2 | XP_015632717.1 | 5 - 6 | 712 | 123 | |
GenBank mRNA | AK121020.1 | - | 5 - 6 | 657 | 123 | |
AY224468.1 | - | 5 - 6 | 529 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
