Assay Details
| Gene Symbol: LOC4332118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776570.2 | XP_015632056.1 | 4 - 5 | 2949 | 120 | |
| GenBank mRNA | AK073981.1 | - | 4 - 5 | 2894 | 120 | |
More Information
Set Membership: |
|

LOC4332118
Genomic Map