Assay Details
| Gene Symbol: LOC4332118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776570.2 | XP_015632056.1 | 7 - 8 | 3488 | 77 | |
| GenBank mRNA | AK073981.1 | - | 7 - 8 | 3433 | 77 | |
More Information
Set Membership: |
|

LOC4332118
Genomic Map