Assay Details
| Gene Symbol: LOC4332255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773462.2 | XP_015628948.1 | 3 - 4 | 486 | 130 | |
| XM_015773463.2 | XP_015628949.1 | 3 - 4 | 487 | 130 | ||
| GenBank mRNA | AK069944.1 | - | 3 - 4 | 474 | 130 | |
More Information
Set Membership: |
|

LOC4332255
Genomic Map