Assay Details
| Gene Symbol: LOC4332255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773462.2 | XP_015628948.1 | 4 - 5 | 549 | 140 | |
| XM_015773463.2 | XP_015628949.1 | 4 - 5 | 550 | 140 | ||
| GenBank mRNA | AK069944.1 | - | 4 - 5 | 537 | 140 | |
More Information
Set Membership: |
|

LOC4332255
Genomic Map