Assay Details
Gene Symbol: LOC4332368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776926.2 | XP_015632412.1 | 11 - 12 | 1518 | 89 | |
XM_015776928.2 | XP_015632414.1 | 10 - 11 | 1325 | 89 | ||
XM_015776929.2 | XP_015632415.1 | 10 - 11 | 1323 | 89 | ||
GenBank mRNA | AK069586.1 | - | 10 - 11 | 1282 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
