Assay Details
| Gene Symbol: LOC4332368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776926.2 | XP_015632412.1 | 10 - 11 | 1397 | 99 | |
| XM_015776928.2 | XP_015632414.1 | 9 - 10 | 1204 | 99 | ||
| XM_015776929.2 | XP_015632415.1 | 9 - 10 | 1202 | 99 | ||
| GenBank mRNA | AK069586.1 | - | 9 - 10 | 1161 | 99 | |
More Information
Set Membership: |
|

LOC4332368
Genomic Map