Assay Details
| Gene Symbol: LOC4332441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775068.2 | XP_015630554.1 | 3 - 4 | 939 | 95 | |
| GenBank mRNA | AB167397.1 | - | 4 - 5 | 805 | 95 | |
| AK071034.1 | - | 3 - 4 | 900 | 95 | ||
More Information
Set Membership: |
|

LOC4332441
Genomic Map