Assay Details
| Gene Symbol: LOC4332622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001360200.1 | NP_001347129.1 | 509 | 90 | ||
| GenBank mRNA | AK070071.1 | - | 2 - 3 | 416 | 90 | |
More Information
Set Membership: |
|

LOC4332622
Genomic Map