Assay Details
Gene Symbol: LOC4332665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775610.2 | XP_015631096.1 | 7 - 8 | 736 | 84 | |
XM_015775611.2 | XP_015631097.1 | 8 - 9 | 863 | 84 | ||
XM_015775612.2 | XP_015631098.1 | 7 - 8 | 1254 | 84 | ||
XM_015775613.2 | XP_015631099.1 | 7 - 8 | 744 | 84 | ||
GenBank mRNA | AK111752.1 | - | 7 - 8 | 1260 | 84 | |
AK111920.1 | - | 4 - 5 | 269 | 84 | ||
AK111929.1 | - | 6 - 7 | 568 | 84 | ||
AY256847.1 | - | 7 - 8 | 689 | 84 | ||
EU703794.1 | - | 6 - 7 | 568 | 84 | ||
EU807984.1 | - | 6 - 7 | 568 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
