Assay Details
Gene Symbol: LOC4332665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775610.2 | XP_015631096.1 | 8 - 9 | 865 | 116 | |
XM_015775611.2 | XP_015631097.1 | 9 - 10 | 992 | 116 | ||
XM_015775612.2 | XP_015631098.1 | 8 - 9 | 1383 | 116 | ||
XM_015775613.2 | XP_015631099.1 | 8 - 9 | 873 | 116 | ||
GenBank mRNA | AK111752.1 | - | 8 - 9 | 1389 | 116 | |
AK111920.1 | - | 5 - 6 | 398 | 116 | ||
AK111929.1 | - | 7 - 8 | 697 | 116 | ||
AY256847.1 | - | 8 - 9 | 818 | 116 | ||
EU703794.1 | - | 7 - 8 | 697 | 116 | ||
EU807984.1 | - | 7 - 8 | 697 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
