Assay Details
Gene Symbol: LOC4332803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772491.2 | XP_015627977.1 | 4 - 5 | 1171 | 121 | |
XM_015772492.2 | XP_015627978.1 | 4 - 5 | 1177 | 121 | ||
GenBank mRNA | AK060603.1 | - | 4 - 5 | 666 | 121 | |
AK102274.1 | - | 3 - 4 | 766 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
