Assay Details
Gene Symbol: LOC4332917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776156.2 | XP_015631642.1 | 1 - 2 | 300 | 65 | |
GenBank mRNA | AK071364.1 | - | 1 - 2 | 271 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776156.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
