Assay Details
| Gene Symbol: LOC4333591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773359.2 | XP_015628845.1 | 2 - 3 | 452 | 94 | |
| GenBank mRNA | AK120336.1 | - | 2 - 3 | 373 | 94 | |
More Information
Set Membership: |
|

LOC4333591
Genomic Map