Assay Details
Gene Symbol: LOC4333920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777503.2 | XP_015632989.1 | 2 | 247 | 93 | |
XM_015777505.2 | XP_015632991.1 | 1 | 459 | 93 | ||
GenBank mRNA | AK108509.1 | - | 1 | 464 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777503.2, XM_015777505.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
