Assay Details
| Gene Symbol: LOC4334045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774318.2 | XP_015629804.1 | 18 - 19 | 11481 | 74 | |
| XM_015774319.2 | XP_015629805.1 | 19 - 20 | 11547 | 74 | ||
| XM_015774320.2 | XP_015629806.1 | 19 - 20 | 11544 | 74 | ||
| GenBank mRNA | AK071866.1 | - | 1 - 2 | 1001 | 74 | |
More Information
Set Membership: |
|

LOC4334045
Genomic Map