Assay Details
| Gene Symbol: LOC4334068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775957.2 | XP_015631443.1 | 1 - 2 | 706 | 78 | |
| GenBank mRNA | AK069015.1 | - | 1 - 2 | 694 | 78 | |
| AK102769.1 | - | 1 - 2 | 671 | 78 | ||
More Information
Set Membership: |
|

LOC4334068
Genomic Map