Assay Details
Gene Symbol: LOC4334399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772852.2 | XP_015628338.1 | 1 - 2 | 542 | 74 | |
GenBank mRNA | AK058812.1 | - | 1 - 2 | 428 | 74 | |
AK061537.1 | - | 1 - 2 | 426 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
