Assay Details
Gene Symbol: LOC4334401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775568.2 | XP_015631054.1 | 1 - 2 | 501 | 150 | |
GenBank mRNA | AK108123.1 | - | 1 - 2 | 412 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
