Assay Details
| Gene Symbol: LOC4334590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772371.2 | XP_015627857.1 | 1 - 2 | 175 | 105 | |
| GenBank mRNA | AK111912.1 | - | 1 - 2 | 165 | 105 | |
| AK112029.1 | - | 1 - 2 | 164 | 105 | ||
| AY576526.1 | - | 1 - 2 | 160 | 105 | ||
| AY579410.1 | - | 1 - 2 | 160 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772371.2 transcripts.
Set Membership: |
|

LOC4334590
Genomic Map