Assay Details
| Gene Symbol: LOC4334590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772371.2 | XP_015627857.1 | 3 - 4 | 349 | 73 | |
| GenBank mRNA | AK111912.1 | - | 3 - 4 | 339 | 73 | |
| AK112029.1 | - | 3 - 4 | 338 | 73 | ||
| AY576526.1 | - | 3 - 4 | 334 | 73 | ||
| AY579410.1 | - | 3 - 4 | 334 | 73 | ||
More Information
Set Membership: |
|

LOC4334590
Genomic Map