Assay Details
Gene Symbol: LOC4334590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772371.2 | XP_015627857.1 | 4 - 5 | 452 | 132 | |
GenBank mRNA | AK111912.1 | - | 4 - 5 | 442 | 132 | |
AK112029.1 | - | 4 - 5 | 441 | 132 | ||
AY576526.1 | - | 4 - 5 | 437 | 132 | ||
AY579410.1 | - | 4 - 5 | 437 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
