Assay Details
| Gene Symbol: LOC4334590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772371.2 | XP_015627857.1 | 6 - 7 | 609 | 107 | |
| GenBank mRNA | AK111912.1 | - | 6 - 7 | 599 | 107 | |
| AK112029.1 | - | 6 - 7 | 598 | 107 | ||
| AY579410.1 | - | 6 - 7 | 594 | 107 | ||
More Information
Set Membership: |
|

LOC4334590
Genomic Map