Assay Details
Gene Symbol: LOC4334590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772371.2 | XP_015627857.1 | 8 - 9 | 765 | 131 | |
GenBank mRNA | AK111912.1 | - | 8 - 9 | 755 | 131 | |
AK112029.1 | - | 8 - 9 | 754 | 131 | ||
AY579410.1 | - | 8 - 9 | 750 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
