Assay Details
Gene Symbol: LOC4334649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774922.1 | XP_015630408.1 | 9 - 10 | 2280 | 61 | |
GenBank mRNA | AK062046.1 | - | 6 - 7 | 1225 | 61 | |
AK065849.1 | - | 10 - 11 | 2227 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
