Assay Details
Gene Symbol: LOC4334649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774922.1 | XP_015630408.1 | 3 - 4 | 1019 | 89 | |
XM_015774923.1 | XP_015630409.1 | 3 - 4 | 943 | 89 | ||
GenBank mRNA | AK065849.1 | - | 4 - 5 | 966 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
