Assay Details
Gene Symbol: LOC4334649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774922.1 | XP_015630408.1 | 4 - 5 | 1113 | 95 | |
XM_015774923.1 | XP_015630409.1 | 4 - 5 | 1037 | 95 | ||
GenBank mRNA | AK062046.1 | - | 1 - 2 | 62 | 95 | |
AK065849.1 | - | 5 - 6 | 1060 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
