Assay Details
Gene Symbol: LOC4334649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774922.1 | XP_015630408.1 | 6 - 7 | 1716 | 122 | |
XM_015774923.1 | XP_015630409.1 | 6 - 7 | 1640 | 122 | ||
GenBank mRNA | AK062046.1 | - | 3 - 4 | 661 | 122 | |
AK065849.1 | - | 7 - 8 | 1663 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
