Assay Details
Gene Symbol: LOC4334649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774922.1 | XP_015630408.1 | 7 - 8 | 1868 | 81 | |
XM_015774923.1 | XP_015630409.1 | 7 - 8 | 1792 | 81 | ||
GenBank mRNA | AK062046.1 | - | 4 - 5 | 813 | 81 | |
AK065849.1 | - | 8 - 9 | 1815 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
