Assay Details
Gene Symbol: LOC4334764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772331.2 | XP_015627817.1 | 1 - 2 | 424 | 50 | |
GenBank mRNA | AK073441.1 | - | 1 - 2 | 431 | 50 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
