Assay Details
Gene Symbol: LOC4334764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772331.2 | XP_015627817.1 | 4 - 5 | 818 | 130 | |
GenBank mRNA | AK073441.1 | - | 4 - 5 | 825 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
