Assay Details
Gene Symbol: LOC4335371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778454.2 | XP_015633940.1 | 1 - 2 | 374 | 59 | |
GenBank mRNA | AK061149.1 | - | 1 - 2 | 322 | 59 |
Gene Symbol: LOC4335106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK240854.1 | - | 326 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
