Assay Details
Gene Symbol: LOC4335371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778454.2 | XP_015633940.1 | 2 - 3 | 494 | 56 | |
GenBank mRNA | AK061149.1 | - | 2 - 3 | 442 | 56 |
Gene Symbol: LOC4335106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK240854.1 | - | 446 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
