Assay Details
| Gene Symbol: LOC4335920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780933.2 | XP_015636419.1 | 1 - 2 | 551 | 70 | |
| GenBank mRNA | AK065524.1 | - | 1 - 2 | 516 | 70 | |
More Information
Set Membership: |
|

LOC4335920
Genomic Map