Assay Details
| Gene Symbol: LOC4335994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780768.2 | XP_015636254.1 | 4 - 5 | 657 | 92 | |
| GenBank mRNA | AK069194.1 | - | 4 - 5 | 659 | 92 | |
| AK100647.1 | - | 4 - 5 | 528 | 92 | ||
More Information
Set Membership: |
|

LOC4335994
Genomic Map